Learn More
Andwin Scientific LTS TIPS LIQUIDATOR 96 LQR-200
SDP
Supplier:  Andwin Scientific 17010645
Encompass_Preferred
The 200µL, Tips in racks are compatible to use with bench-top pipettor with LTS™. The tips have a volume range of 5 µL – 200 µL. This product follows the standard SBS footprint. There are 960 tips in 10 racks of 96.
Catalog No. NC0909707
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.