Learn More
Teledyne ISCO, Inc Ls Class Single Piston Pump With Belt Drive And Shaped Cam; 0.0 - 40.0 Ml/Min. ; 1,600 Ps
Supplier:  Teledyne ISCO, Inc LS040SRX
Encompass
LS Class Single Piston Pump with Belt Drive and Shaped Cam; 0.0 - 40.0 mL/min. ; 1,600 psi; Stainless Steel Fluid Path; Automatic Piston Wash; with Pulse Dampener; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/16"""" tubing
Catalog No. 50-211-9950
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.