Learn More
Teledyne ISCO, Inc Ls Class Single Piston Pump With Belt Drive And Shaped Cam; 0.000 - 5.000 Ml/Min.; 6,000 Psi
Supplier: Teledyne ISCO, Inc LS005PRX
LS Class Single Piston Pump with Belt Drive and Shaped Cam; 0.000 - 5.000 mL/min.; 6,000 psi; PEEK Fluid Path; Automatic Piston Wash; with Pulse Dampener; Standard Seal (Flouropolymer energizer); Fitting package for 1/8"""" and 1/16"""" tubing