Learn More
New England Biolabs, Inc. Low Range ssRNA Ladder – 100 gel lanes
SDP
Supplier:  New England Biolabs, Inc. N0364S
Encompass
The Low Range ssRNA Ladder is a set of 6 RNA molecules produced by in vitro transcription of a mixture of 6 linear DNA templates. The ladder sizes are: 1000, 500, 300, 150, 80 and 50 bases. The 300 base fragment is at double intensity to serve as a reference band. This ladder is suitable for use as an ssRNA size standard on denaturing polyacrylamide-urea gels, and on denaturing or native agarose gels.
Catalog No. 50-994-976
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.