Learn More
Electron Microscopy Sciences Low-Intensity UV Lamp, 220 V
Supplier: Electron Microscopy Sciences 72423
This 6-watt UV lamp meets most laboratory requirements; such as, bacterial identification, fluorchemistry, specimen staining, titration processes, photochemical reactions, aflatoxin detection, and curing UV cements. Constructed from Cyolac plastic, with a longwave emission of 365nm and an intensity at 6" (15.2cm) of 750 um/cm squared. The bulb has a rated life of 6,000 hours. It can be used as a hand-held light or it may br mounted on a optional stand.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.