Learn More
Sarstedt Inc LOW DNA BIND TUBE 1.5ML 200/PK
Supplier:  Sarstedt Inc 72.706.700
Encompass_Preferred
SafeSeal reaction tube DNA low binding, 1.5 ml, PP, with attached lid with retaining cams, with moulded graduation and frosted writing space, certified PCR Performance Tested, 50 pcs./minigrip bag
Catalog No. NC0964895
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.