Learn More
Electron Microscopy Sciences Maxtaform London Honeycomb H 6, 200 mesh 200 Mesh, 235 µm Pitch
Supplier:  Electron Microscopy Sciences LH200CU
Encompass_Preferred
Maxtaform grids with reference patterns are of the highest consistent quality.
Material: 200 Mesh
Pitch: 235 µm
Catalog No. 50-269-77
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.