Learn More
Andwin Scientific LIQUI-NOX, GALLON 4/CS
Supplier: Andwin Scientific 21837027
The Liqui-Nox is a specialized cleaning detergent for laboratory equipments. With dilution ratio of 1:100, this safe, odorless, biodegradable, and concentrated cleaning product can be rinsed without any interfering residue. This product is completely soluble in either hard or soft water. It is a basic cleaner with a pH of 8.5. This is sold in a case of 4 of 1 gallon.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.