Learn More
Neogen Diagnostics LIPOXIN A4 ELISA - 96 WELL
Supplier:  Neogen Diagnostics 407010
Encompass_Preferred
Lipoxin A4 ELISA is used for the quantitative analysis of Lipoxin A4 levels in biological fluid. Lipoxin A4 (LXA4) is a biologically active lipoxygenase interaction product derived from arachidonic acid. Arachidonic acid is first oxygenated by 15-lipoxygenase to form 15-HETE which is then converted by 5-lipoxygenase and epoxide hydrase to generate LXA4. LXA4 stimulates leukocyte chemotaxis without aggregation and inhibits natural killer cell cytotoxicity. It also provokes contraction of parenchymal strips and stimulates microvascular changes. Recent findings indicate that LXA4 inhibits leukocyte-dependent inflammation. Determination of LXA4 levels may provide new understanding of the role of LXA4 in basic cellular reactions and in pathophysiology of inflammation and other disease processes. Store at -20°C for lyophilized conjugate, 2-8°C for all other kit components.
Catalog No. NC2215600
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.