Learn More
Neuromics, Inc Lipid Depleted Fetal Bovine Serum, 500 ml has reduced concentrations of lipids. Best for use in research where a high concentration of lipid may interfere.
Supplier: Neuromics, Inc FBS005L
Our FBS is manufactured in a fully integrated environment from raw material collection to finished product. It is produced to the highest standards in order to reduce risk and product variability and to ensure product safety and performance.Our Lipid Depleted FBS features: Reduced concentration of lipids, Triple 0.1 μm Filtered, and Free of Virus and Mycoplasma.Lipid Depleted FBS reduces the concentration of lipids (free fatty acids, etc.). Best used for research applications where the normal concentration of lipids may interfere.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.