Learn More
World Precision Instrument Linear Stimulus Isolator-A395RC
SDP
Supplier:  World Precision Instrument A395RC
Encompass_Preferred
Model A395 generates a user-defined output current of wave shape; DC, AC, pulse and combinations. Battery operated, pho­to­elec­tri­cal­ly-isolated from the input voltage drive, the instrument regen­er­ates out­put currents which are linearly proportional to the analog voltage wave­forms pro­vid­ed by your D/A con­vert­er or sig­nal generator.
Catalog No. 50-253-8936
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.