Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lightning-Link Rapid Rhodamine Antibody Labeling Kit
Label antibodies, proteins, peptides and other biomolecules with rhodamine, using simple procedure. Set up conjugations in seconds, and use them within minutes.
$546.00 - $1153.00
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
---|---|
Product Type | Antibody Labeling Kit |
Alert:
For Research Use Only
Description
- No desalting or dialysis steps
- Lightning-link Rapid technology can be used to label small quantities of antibody with 100% recovery
- Rhodamine used in kit has absorption peaks at 520 and 544nm
- Maximum emission is obtained when the rhodamine is excited at 544nm
Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
Antibody Labeling Kit |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title