Learn More
Novus Biologicals™ Lightning-Link (R) Rapid DyLight 800 Antibody Labeling Kit
SDP
Antibody labeling kit
Supplier:  Novus Biologicals™ 3290010
 View more versions of this product
Catalog No. 32-900-10
Explore all special offers
Add to Cart
Description
Description
DyLight 800 provides near infrared fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 770nm, high fluorescence at 794nm (extinction coefficient 2.7 x10&superscript_begin;5&superscript_end;cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.
Order Info
Quantity
Specifications
Specifications
DyLight 800
Store at -20°C. Avoid freeze-thaw cycles.
300 μg
Rapid DyLight 800 Antibody Labeling Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.