Learn More
Electron Microscopy Sciences Leur Bone Rongeur 3 mm Blade
Supplier: Electron Microscopy Sciences 7798301
Bone Rongeur, curved. Delicate. The jaws width is 3 mm. and it is 6"long. Made from stainless steel with a polished finish. Curved, delicate, the jaws width is 3mm and it is 6"long.