Learn More
STEMCELL Technologies Human Recombinant LIF, ACF, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 78149.1
Encompass_Preferred
Use leukemia inhibitory factor (LIF) to regulate a variety of developmental functions. LIF is an interleukin 6 class cytokine that regulates proliferation, invasion, and differentiation of trophoblasts following blastocyst attachment. Upon binding of LIF to LIF receptor (LIFR), LIFR associates with gp130 and activates JAK/STAT and MAPK signaling (Suman et al. JAK-STAT, 2014). LIFR activation of STAT3 is essential for maintaining the mouse embryonic stem cell (ESC) phenotype (Niwa et al. Genes Dev, 1998). Human LIF can be used for the maintenance of mouse ESCs (Dahéron et al. Stem Cells, 2004). Human recombinant LIF from STEMCELL is animal component-free and comes lyophilized with ≥ 95% purity; EC50 ≤ 0.2 ng/mL; endotoxin levels verified ≤ 1 EU/μg protein. Alternative names: CDF, Cholinergic differentiation factor, D Factor, DIA, Differentiation-inducing factor, Differentiation inhibitory activity, Emfilermin, Hepatocyte-stimulating factor III, Leukemia inhibitory factor, MLPLI.
Catalog No. 50-226-9687
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.