Learn More
Medchemexpress LLC HY-B2235 100mg Medchemexpress, Lecithin CAS:8002-43-5 Purity:>98%
Supplier: Medchemexpress LLC HYB2235100MG
Medchemexpress, HY-B2235 100mg Lecithin CAS:8002-43-5 Lecithin is regarded as a safe, conventional phospholipid source. Phospholipids are reported to alter the fatty acid composition and microstructure of the membranes in animal cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.