Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Lanthanum Nitrate, Hexahydrate, Reagent 50 GR

Supplier:  Electron Microscopy Sciences 17300

Encompass_Preferred

CAS #10277-43-7 La(NO₃)₃·6H₂₀ F.W. 433.03
Assay: Min 99.9%
Melting point: 40 °C (104 °F)

A metal stain, intracellular tracer. Lane, N,J. and Treherne, J.E. (1970). Lanthanum staining of neurotubules in axons from cockroach ganglia. J. Cell Sci., 7:217 and Lane, N.J. (1972) Fine structure of a lipidopteran nervous system and its accessibility to peroxidase and lanthanum. Z. Zellforsch., 131:205

Catalog No. 50-268-76


Explore all special offers
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.