Learn More
New England Biolabs, Inc. LAMP Fluorescent Dye - 250 ul
SDP
Supplier:  New England Biolabs, Inc. B1700S
Encompass
The LAMP fluorescent dye is an intercalating dye that can be used to track a LAMP reaction in real-time by fluorescence measurement. It is provided at 50X concentration and is sufficient for at least 500 LAMP reactions (0.5 ul dye per 25 ul LAMP reaction). The SYBR/FAM channel of common real-time fluorimeters can be used to read the dye.
Catalog No. 50-206-8319
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.