Learn More
New England Biolabs, Inc. Lambda DNA – 1250 µg
SDP
Supplier:  New England Biolabs, Inc. N3011L
Encompass
Lambda DNA is a commonly used DNA substrate. It is duplex DNA is isolated from bacteriophage lambda (cI857ind 1 Sam 7). Lambda DNA is 48,502 base pairs in length.
Catalog No. 50-811-746
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.