Learn More
Laboratory Fortifying Stock Solution 1, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$670.00 - $1127.00
Specifications
Chemical Name or Material | LFS Solution 1 without Mercury |
---|---|
Packaging | Triple-leached LDPE bottle |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Analyte or Component Names | 50μg/mL: P; 25μg/mL: Al,As,B |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
LFS1100N
|
SPEX CertiPrep
LFS1100N |
125 mL | Triple-leached LDPE bottle |
Each for $670.00
|
|
||||
LFS1500N
|
SPEX CertiPrep
LFS1500N |
500 mL | Triple-leached LDPE bottle |
Each for $1,127.00
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
P | 50μg/mL |
Al | 25μg/mL |
As | 25μg/mL |
B | 25μg/mL |
Ba | 25μg/mL |
Cr | 25μg/mL |
Cu | 25μg/mL |
Fe | 25μg/mL |
Li | 25μg/mL |
Mn | 25μg/mL |
Ni | 25μg/mL |
Pb | 25μg/mL |
Sb | 25μg/mL |
Se | 25μg/mL |
SiO2 | 25μg/mL |
Sr | 25μg/mL |
Tl | 25μg/mL |
Zn | 25μg/mL |
Cd | 10μg/mL |
Co | 10μg/mL |
Mo | 10μg/mL |
Sn | 10μg/mL |
V | 10μg/mL |
Be | 5μg/mL |
Ag | 2.5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
LFS Solution 1 without Mercury | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
50μg/mL: P; 25μg/mL: Al,As,B |
Triple-leached LDPE bottle | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
5% HNO3/tr Tart. Acid/tr HF |
Safety and Handling
Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.