Learn More
Enzo Life Sciences L-Thiocitrulline . dihydrochloride (50mg). CAS: 212051-53-1
Supplier: Enzo Life Sciences ALX106013M050
Inhibitor of neuronal (nNOS; NOS I; Ki=60nM) and inducible (iNOS; NOS II) nitric oxide synthase. Formula: C6H13N3O2S . 2HCl. MW: 191.3 . 72.9. Purity: ≥97% (TLC). Appearance: White solid. Solubility: Soluble in water or DMSO. Long Term Storage: +4°C.