Learn More
Bellco Glass, Inc. LShaped Cell Spreader is ideal for evenly spreading bacterial cultures angled tip prevents damaging agar surface.Specifications Color Green Length 145mm Case Quantity 500
Supplier: Bellco Glass, Inc. 229616000
LShaped Cell Spreader is ideal for evenly spreading bacterial cultures angled tip prevents damaging agar surface.Specifications Color Green Length 145mm Case Quantity 500