Learn More
Pfanstiehl Inc L-Glutamine, High Purity, Low Endotoxin, Low Metals, USP, JP, ChP (G-137) - 100G, GMP, Multicompendial -CAS: 56-85-9
Supplier:  Pfanstiehl Inc G137RD
Encompass_Preferred
L-Glutamine is an essential amino acid involved in numerous physiological processes within mammalian cells. In upstream cell culture protein processes, L-Glutamine plays a pivotal role in supporting cell growth, viability, and protein production.
Catalog No. NC2953523
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.