Learn More
Bellco Glass, Inc. Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Can be used with series 500 contro
SDP
Supplier:  Bellco Glass, Inc. 196488080
Encompass_Preferred
Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Can be used with series 500 contro
Catalog No. 50-194-4880
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.