Learn More
Enzo Life Sciences L-Ergothioneine (50mg). CAS: 497-30-3
SDP
Supplier:  Enzo Life Sciences BMLFR1110050
Encompass_Preferred
Naturally occuring ubiquitous antioxidant which is found in many cells at concentrations up to 2 mM. Peroxynitrate ONOO- scavenger. Protects human neuronal hybridoma cell line (N-18-RE-105) from ONOO--induced oxidative damage. Purity: ≥98% (TLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0833
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.