Learn More
Enzo Life Sciences L-Cycloserine (100mg). CAS: 339-72-0
SDP
Supplier:  Enzo Life Sciences ALX106037M100
Encompass_Preferred
Ketosphinganine synthase inhibitor. Excitatory amino acid. Inhibitor of ketosphinganine synthase (serine-palmitoyl-CoA transferase, EC 2.3.1.50), leading to blockade of sphingosine biosynthesis. Partial agonist at the glycine modulatory site of the NMDA receptor. Alternative name: S(-)-4-Amino-3-isoxazolidinone. Formula: C3H6N2O2. MW: 102.1. MI: 14: 2751. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water. Use/Stability: Unstable in neutral or acidic solutions. Long Term Storage: -20°C. Handling: Hygroscopic.
Catalog No. 50-200-7744
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.