Learn More
Hach Company Phenols Reagent Set (No Air Shipments)
Supplier:  Hach Company 2243900
Encompass_Preferred
For determination of Phenols by the 4-Aminoantipyrine method. Hach Method 8047. USEPA accepted for reporting wastewater analyses. Range: 0.002 - 0.200 mg/L Reagent Set includes: Hardness 1 Buffer Solution, Chloroform, Phenol Reagent Powder Pillows, and Phenol 2 Reagent Powder Pillows for 100 tests. For air freight shipments, order 2243901
Catalog No. 50-199-5623
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.