Learn More
Hach Company Probe Cable Depth Markers for Rugged IntelliCAL Probes
Supplier: Hach Company 5828610
Taking readings at specific depths with rugged outdoor electrodes is a snap with these depth markers. Visually identify the depth of your electrode by attaching depth marker securely on cable at points to meet your needs. Five markers per package.Note: For use with rugged outdoor electrodes only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.