Learn More
Hach Company m-ColiBlue24 Broth, Bulk Glass Bottles, Economy Kit, 200 test
Supplier:  Hach Company 2779201
Encompass_Preferred
Use m-ColiBlue24 Broth to accurately monitor and evaluate total coliform and E. coli in drinking water; wastewater; bottled water; beverages; surface, ground, and well water; and ultrapure, chemical processing and pharmaceutical processing waters.In 24 hours, Hach's m-ColiBlue24 Broth give you simultaneous results for total coliforms and E. coli. With ready-to-use m-ColiBlue24 Broth, you can enumerate total coliforms and E. coli on one petri dish. Differentiate colonies easily - red and blue indicate total coliforms and blue specifies E. coli. Minimize background growth of non-coliform bacteria.MCB24 Economy Kits include: 200 PALL Metricel GN-6 Membranes (0.45 µm; sterile; indiv. packed), 200 PALL 50 mm petri dishes w/nutrient pad, 4x100 mL Glass Bottles (200 test).
Catalog No. 50-199-6672
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.