Learn More
Hach Company HQ440D High Performance Water Laboratory pH Meter Package for Extended Temperature Range Measurements with Fast Response RedRod PHC705 Glass Refill Electrode
Supplier:  Hach Company 8506050
Encompass_Preferred
Designed for water quality experts, the Hach HQ440D Laboratory Multi-Meter is an advanced laboratory meter that takes the guesswork out of measurements. HQD digital meters connect with smart Intellical electrodes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced HQD Laboratory Meter is designed to withstand years of continuous lab use. HQD bench meters feature a simple user interface that does not require manuals or training to operate. The Intellical PHC705 is a digital, combination glass-body electrode with a refillable KCl reference and built-in temperature sensor. The PHC705 utilizes Red Rod technology to deliver exceptionally fast response times and stable, accurate measurements. It has a porous pin junction and is ideal for measuring pH in a variety of laboratory applications.
Catalog No. 50-199-7240
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.