Learn More
Hach Company HQ4200 Portable Multi-Meter with Gel pH and Dissolved Oxygen electrode, 5 m Rugged Cables
Supplier:  Hach Company LEV015.53.42006
Encompass_Preferred
HQ4200 Portable pH/ORP/EC/TDS/DO Meter, 2 channel for waste water plant testing with IntelliCAL PHC101 pH electrode and LDO101 DO probe, 5 m Rugged Cables. Field accessories included.
Catalog No. 50-206-2973
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.