Learn More
Sino Biological KLH PAb 100ug (Sino Biological)
Supplier:  Sino Biological 12133T16B100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant KLH (Keyhole Limpet Hemocyanin). KLH specific IgG was purified by KLH affinity chromatography and then biotinylated.
Catalog No. 50-237-1587
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.