Learn More
Chemglass Life Sciences Kit, Vial, 2.0mL, Amber with Spot, 12 x32mm, with 0.30mL Fused Insert, .040" PTFE/Silicone Septa
SDP
Supplier:  Chemglass Life Sciences CV51811232
Encompass_Preferred
Kit, Vial, 2.0mL, Amber with Spot, 12 x32mm, with 0.30mL Fused Insert, .040" PTFE/Silicone Septa. These convenience packs come with 100 bonded closures and vials packed seperately. Closures with bonded septa are not attached.
Catalog No. 50-153-2638
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.