Learn More
Chemglass Life Sciences Kit, Vial, 2.0mL, Clear with Spot, 12 x32mm, with 0.30mL Fused Insert, .040" PTFE/Silicone Septa with Slit
Supplier: Chemglass Life Sciences CV51821232
Kit, Vial, 2.0mL, Clear with Spot, 12 x32mm, with 0.30mL Fused Insert, .040" PTFE/Silicone Septa with Slit. These convenience packs come with 100 bonded closures and vials packed separately. Closures with bonded septa are not attached.