Learn More
Eurofins Discoverx KILR Retroparticles for Suspension Cells (Hygromycin B), 4 vials x 1.0. (97-0006)
Supplier:  Eurofins Discoverx 970006
Encompass_Preferred
The KILR Retroparticles for Suspension Cells (Hygromycin B) are MMLV particles designed to transduce suspension cells. This allows production of ProLabel-tagged, hygromycin B-resistant target cells of interest for use in cytotoxicity assays involving detection of cell lysis. This product uses EFC technology. The retroparticles mix can be used to transduce target cells with a ProLabel-tagged housekeeping gene. Target cell death in a cytotoxicity assay will release ProLabel to the media, where EA contained in the detection reagent will produce EFC signal. These retroparticles ship as 4 vials of cryopreserved retroparticles. For detailed product information, User Manual and datasheet, visit www.discoverx.com/products/virus/kilr/retroparticles-for-suspension-cells-(hygr-97-0006.
Catalog No. 50-200-4591
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.