Learn More
Neogen Diagnostics K-Blue Select TMB Substrate, 500 ML, HRP, Microwell, Colorimetric
Supplier:  Neogen Diagnostics 382176
Encompass_Preferred
K-Blue Select is a one-bottle stabilized chromogenic substrate for use with horseradish peroxidase immunoassays. It contains both 3,3’,5,5’ tetramethylbenzidine (TMB) and hydrogen peroxide (H2O2) in a one-bottle format with long term stability. K-Blue Select offers mid-level activity range. K-Blue Select is compliant with Annex XVII for REACH and contains no hazardous substances at or above published reporting levels. K-Blue Select develops a deep blue color in the presence of peroxidase labeled conjugate. K-Blue Select has been optimized for use with microwell applications, especially ELISA. It is not applicable for use with assays requiring a precipitating substrate. Stable for a minimum of 24-months when stored at 2-8C.
Catalog No. NC2215581
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.