Learn More
Neogen Diagnostics K-Blue Luminescent ONE Substrate, 500 ML, HRP, Microwell, Chemiluminescent
Supplier:  Neogen Diagnostics 384176
Encompass_Preferred
K-Blue Luminescent ONE is a stabilized luminescent substrate for use with horseradish peroxidase immunoassays. It contains both luminol and hydrogen peroxide (H2O2) in a one-bottle format with long-term stability, high sensitivity, and excellent signal to noise. Maximal light emission occurs at 430 nm in the presence of peroxidase labeled conjugate. K-Blue Luminescent ONE is compliant with Annex XVII for REACH and contains no hazardous substances at or above published reporting levels. Stable for a minimum of 12-months when stored at 2-8C.
Catalog No. NC2215605
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.