Learn More
World Precision Instrument Joystick Controlled Manual Micromanipulator-MMJR
Supplier: World Precision Instrument MMJR
Specially adapted for use with the Nanoliter Injector for oocyte injection and similar applications, this joystick-controlled micromanipulator allows an easy “steering” motion that translates normal hand movement into smooth sub-millimeter shifts. Viewed microscopically, movement of the tool-tip corresponds naturally to hand movement, so accurate resolution is intuitive and quick. All fine adjustment can be controlled by the joystick. Pivoting forward, backward or laterally gives precise x-y adjustment. For added convenience, a separate coarse control lever is also provided for quick raising and lowering. A stop screw, which is set once resolution is achieved, eliminates refocusing and streamlines repetitive work by guiding the tip to its previous focusing plane.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.