Learn More
Chemglass Life Sciences Support, Chem-Jack, Lattice Mount, fits 1/2in OD Rods
SDP
Supplier:  Chemglass Life Sciences CG305301
Encompass_Preferred
Support, Chem-Jack, Lattice Mount, fits 1/2in OD Rods. Features:Only support jack on the market that attaches directly to a 1/2" diameter vertical lab frame or lattice rod.Heavy gauge aluminum construction provides excellent rigidity and support.Fabricated to precise tolerances allowing smooth vertical adjustment.Optional stainless steel top plate allowing hotplate stirrers to be positioned on the Chem-Jack.Raises and lowers loads up to 15lbs through a vertical distance of 2.75" to a maximum of 7.75", Closes to a height of 2.75". Top plate is 6.25" W x 7.5" L.Chem-Jack is the only support jack on the market which attaches directly to a 1/2" diameter vertical lab-frame rod. The scissor-style jack is constructed of heavy gauge, non-sparking, anodized aluminum and has a thick silicone pad to prevent apparatus from slipping. The rear of the jack has an open "V" mount permitting secure attachment anywhere on the 1/2" diameter vertical rod.
Catalog No. CG305301
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.