Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropanol, 99%, Vion Biosciences

Catalog No. S02494 Shop All Aldon Products
Click to view available options
Quantity:
1 L
3.8 L
4 x 1 gal.

This 99% isopropanol is an excellent alternative to ethanol as a dehydrant and during paraffinization as an substitute for solvents such as chloroform, xylene, and benzene.

  • This 99% isopropanol is an excellent alternative to ethanol as a dehydrant and during paraffinization as an substitute for solvents such as chloroform, xylene, and benzene.

Chemical Identifiers

Molecular Formula (CH3)2CHOH
Molecular Weight (g/mol) 60.1 g/mol
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym 2-propanol, isopropanol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH Data not available.
Melting Point -90°C
Color Colorless
CAS Max % 100%
Appearance Clear, colorless liquid.
Decomposition Information Data not available.
Density 0.786 to 0.790 (at 20°C)
Boiling Point 82°C
Quantity 4 x 1 gal.
Explosion Limit 2%
CAS 67-63-0
Flash Point 12°C
Identification Isopropanol
Ignition Point 399°C
Infrared Spectrum Yes
Odor Aromatic odor.
Packaging 1 Gallon F Style UN Bottle
Physical Form Liquid
Product Line Vion Histology
Assay Percent Range 100%
Specific Gravity 0.0786 to 0.790 (at 20°C)
Linear Formula (CH3)2CHOH
Vapor Pressure 33 mm (at 20°C)
UN Number UN1219
Water 0%
Concentration 99%
Formulation 95% Isopropanol
Solubility Information Complete in water.
Formula Weight 60.1 g/mol
Percent Purity 99%
Grade Histological
Concentration or Composition (by Analyte or Components) 99% Isopropanol
Chemical Name or Material Isopropanol
Show More Show Less
Hazard Category Causes serious eye irritation.
Hazard Statement May cause drowsiness or dizziness.
ShelfLife 3 Years
DOTInformation UN1219, Isopropanol, 3(II), Limited quantity ≤ 1 L
EINECSNumber 200-661-7
RTECSNumber NT8050000
Recommended Storage Store in a cool, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.