Learn More
Glass Expansion IsoMist XR Kit with Glass Spray Chamber for Thermo 6000/7000 Radial ICP-OES
Supplier: Glass Expansion KT1021XR
IsoMist XR Kit for Thermo 6000/7000 Radial Series. Compatible with Thermo 6000/7000 Radial Series ICP-OES. This IsoMist XS is programmable and temperature-controlled, with a glass cyclonic spray chamber now featuring an improved thermodynamic design that provides an extended temperature range of -25C to +80C. The temperature is settable in 1C increments and can maintain accuracy of + or 0.1C. Proven performance results are, improved long-term stability and precision, no dilution for volatile organic solvents, reduced oxide interferences (ICP-MS), enhanced sensitivity and accuracy, lower detection limits, faster washout, and reduced hazardous waste/disposal fees.The IsoMist XS is also available with a quartz or PFA HF-resistant spray chamber.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.