Learn More
Volu Sol IronView Stain Kit, 8 oz / 250 mL, Volu-Sol™
SDP
Supplier:  Volu Sol VNF250
Encompass_Preferred
Volu-Sol's IronView Stain Kit utilizes a Prussian Blue method similar to Gomori's for the demonstration of ferric (Fe3+) iron in tissues. This kit produces superb results while utilizing lower concentrations of potentially hazardous chemicals. Ferric Iron appears blue and nuclei red, results are present on a pink background counterstained by Nuclear Fast Red.
Catalog No. 50-169-4712
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.