Learn More
Andwin Scientific IPA 70% ST TRGR SP BTL 12/CS
SDP
Supplier:  Andwin Scientific TX3270
Encompass_Preferred
The Texwipe's Sterile 70% Isopropanol (IPA) is composed of 70% by volume USP-grade Isopropanol (isopropyl alcohol) and 30% USP-purified water. This product guarantees sterility as the solution has been 0.2 submicron filtered, filled into cleaned containers, double-bagged, and gamma-irradiated. It is packaged in a 16 oz. trigger spray bottle and is sold in a case of 12.
Catalog No. NC9093906
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.