Learn More
Regis Technologies Inc Ion Pairing Reagents and Buffers, Regis, ION PAIR CONCENTRATE Q12 (100 mL)
SDP
Supplier:  Regis Technologies Inc 1404031200
Encompass_Preferred
Q-Series Ion Pair Concentrates (For Anions) are Regis Quaternary Amines, Bulk Ion Pair Reagents (for Anions). The Q-Series is comprised of quaternary alkyltriethylamines that can be used for the resolution of negatively charged species. This unique set of cationic reagents was developed to complement the Sulfonate Series (S-Series) and is exclusively manufactured by Regis. The Quaternary Alkyltriethylamines are available as premixed 0.5 M solutions of alkyl amines. When diluted to 1 L with HPLC-grade water, a 10 mL bottle forms a 0.005 M buffered solution.
Catalog No. 50-442-958
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.