Learn More
Invitrogen™ Platinum™ II Hot-Start PCR Master Mix (2X)
Invitrogen™ Hot-Start PCR Master Mix for fast and easy PCR
Supplier: Invitrogen™ 14000011
Description
Invitrogen™ Platinum™ Taq II Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq, and leading hot-start technology.
- Innovative buffer—enables universal primer annealing protocol by isostabilizing primer-template duplex structures
- Engineered Taq—confers fast cycling and tolerance to common inhibitors
- Platinum hot-start technology—ensures superior specificity, sensitivity, and yields; allows for room temperature reaction setup
- Optional green buffer—reduces pipetting error with direct gel loading
Additional:
- Antibody-mediated hot-start
- Universal annealing protocool
- Inhibitor resistance
Applications:
- Genotyping
- High-throughput PCR
- GC-rich templates
- Fast PCR
- Amplification of samples with suboptimal purity
- Direct PCR with blood
- PCR from DNA extracted from FFPE samples
- Colony PCR
- PCR from bacterial 16S rRNA genes
Specifications
GC-rich amplification | |
3' A | |
2X Master mix | |
Target length: Up to 5kb | |
PCR Master mix; GC enhancer; Water | |
Store at -20°C |
Genotyping, PCR | |
1X | |
20 reactions | |
5' to 3' | |
No direct loading |
Safety and Handling
ShelfLife : 24 months
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For research use only