Learn More
Chemglass Life Sciences Internal Bushing, #7, PTFE, 1/4" NPT, #008 Viton O-Ring
Supplier: Chemglass Life Sciences CG36422
Internal Bushing, #7, PTFE, 1/4" NPT, #008 Viton O-Ring. New front seal PTFE bushing with female NPT for use with CG-360 internal threads. Supplied complete with Viton o-ring of the listed size. See CG-305 for Viton and CG-309 for Perfluoro replacement o-rings.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.