Learn More
Saf T Pak Inc STP-320R - Category B Insulated Shipping System (UN 3373), Frozen w/ Reusable Secondary Pressure Vessel
Supplier: Saf T Pak Inc STP320R
STP-320R Category B, Insulated Shipping System with Reusable Secondary Pressure Vessel, 1.25L Polypropylene Secondary Pressure Vessel. Validated with Dry Ice, Gel Packs and Saf-T-Temp Paks. Outer Box – OD 15.375 x 13.675 x 12.75in. pre-printed with Biological Substance, Category B, UN 3373 and Exempt Human / Animal Specimen Markings, Insulated Chest – ID 12 x 10.25 x 8in., STP-111 Inner Box – OD 6.5 x 6.5 x 8in., 4/Case
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.