Learn More
Eurofins Discoverx INSL 5 (Mouse), 20 µg. (92-1228)
Supplier:  Eurofins Discoverx 921228
Encompass_Preferred
Insulin-like peptide 5 (INSL5) is a heterodimer of the INSL5A and B peptide chains, and a member of the insulin/relaxin superfamily that is a potent agonist for the G protein-coupled receptor RXFP4. Possible roles include gut contractility, thymic development and regulation, food intake, and glucose homeostasis. Provided as 200 μg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9751
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.