Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Inserts for 2 mL Large Opening Vials
Insert fits 12 x 32 mm vials with large opening (6 mm)
Supplier: MilliporeSigma™ Supelco™ Z2918031PAK
Description
- Glass conical inserts with polymer feet are used to contain standard mixtures for various experimental purposes.
Specifications
6 mm | |
0.3 mL | |
30 mm |
Glass Insert (Conical with Polyspring) | |
100 | |
12 x 32 mm Vials with Large Opening (6 mm) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction