Learn More
Chemglass Life Sciences Polypropylene Insert, 0.25mL, BM with Bottom Spring
SDP
Supplier:  Chemglass Life Sciences CV14250631
Encompass_Preferred
Polypropylene Insert, 0.25mL, BM with Bottom Spring. Due to the conical design of the snap plug, limited volume inserts used in the shell vial stay centered and straight allowing for easy sample removal.
Catalog No. 50-230-846
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.